Assay Details
Target Gene Details
Entrez Gene ID: | 253782 |
Gene Name: | ceramide synthase 6 |
Gene Aliases: |
CERS5, LASS6 |
Location: |
Chr.2:168456249-168775134 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERS6 | NM_001256126.1 | 11 | 6211 | NP_001243055.1 |
| NM_203463.2 | 10 | 6187 | NP_982288.1 | |
| XM_005246440.4 | 8 | 7502 | XP_005246497.1 | |
| XM_017003749.1 | 8 | 6362 | XP_016859238.1 | |
| AK023042.1 | 2 | 1765 | ||
| AK024891.1 | 1 | 1746 | ||
| AK123214.1 | 1 | 2073 | ||
| AL080192.1 | 1 | 1021 | ||
| BM968545.1 |
Target Gene Details
Entrez Gene ID: | 100861402 |
Gene Name: | CERS6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:168771951-168786429 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERS6-AS1 | NR_045786.1 | 4 | 442 | |
| NR_045787.1 | 3 | 349 | ||
| DB072723.1 | 4 | 442 | ||
| DB077980.1 | 3 | 349 |
More Information
Set Membership: |
|

Genomic Map