Assay Details
Target Gene Details
Entrez Gene ID: | 3699 |
Gene Name: | inter-alpha-trypsin inhibitor heavy chain 3 |
Gene Aliases: |
H3P, ITI-HC3, SHAP |
Location: |
Chr.3:52794768-52809009 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITIH3 | NM_002217.3 | NP_002208.3 | ||
| XM_005265105.4 | XP_005265162.1 | |||
| XM_017006356.1 | XP_016861845.1 | |||
| AK222757.1 | BAD96477.1 | |||
| AK223483.1 | BAD97203.1 | |||
| AK298450.1 | ||||
| BC107604.1 | AAI07605.1 | |||
| BC107605.1 | AAI07606.1 | |||
| BC107814.1 | AAI07815.1 | |||
| BX448552.2 | ||||
| BX495022.1 | ||||
| X67055.1 | CAA47439.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508928 | Chr.3:52769883 - 52888575 on Build GRCh38 | Insertion |
|
| nsv1010761 | Chr.3:52770531 - 52817242 on Build GRCh38 | Gain |
|
| esv2762328 | Chr.3:52740868 - 52968134 on Build GRCh38 | Loss |
|
| nsv834698 | Chr.3:52715967 - 52894203 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map