Assay Details
Target Gene Details
Entrez Gene ID: | 51256 |
Gene Name: | TBC1 domain family member 7 |
Gene Aliases: |
MGCPH, PIG51, TBC7 |
Location: |
Chr.6:13304951-13328583 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D7 | NM_001143964.3 | 5 | 741 | NP_001137436.1 |
| NM_001143965.3 | 5 | 601 | NP_001137437.1 | |
| NM_001143966.3 | 4 | 520 | NP_001137438.1 | |
| NM_001258457.2 | NP_001245386.1 | |||
| NM_001318805.1 | 5 | 658 | NP_001305734.1 | |
| NM_016495.5 | 5 | 761 | NP_057579.1 | |
| AB449888.1 | 4 | 465 | ||
| AF151073.1 | 5 | 558 | AAF36159.1 | |
| AK223445.1 | 5 | 638 | BAD97165.1 | |
| AY542308.1 | AAT08177.1 | |||
| AY826820.1 | 4 | 446 | ||
| BC007054.1 | 5 | 562 | AAH07054.1 | |
| BC050465.1 | 4 | 462 | AAH50465.1 | |
| BI914228.1 | ||||
| BP319580.1 | 5 | 532 | ||
| BX354770.2 | 5 | 598 | ||
| DA169008.1 |
More Information
Set Membership: |
|

Genomic Map