Assay Details
Target Gene Details
Entrez Gene ID: | 286365 |
Gene Name: | olfactory receptor family 13 subfamily D member 1 |
Gene Aliases: |
OR9-15 |
Location: |
Chr.9:104694422-104695462 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR13D1 | NM_001004484.1 | 1 | 697 | NP_001004484.1 |
| BC140916.1 | 1 | 740 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891706 | Chr.9:104480016 - 105304573 on Build GRCh38 | Gain |
|
| dgv2175e212 | Chr.9:104694039 - 104697790 on Build GRCh38 | Loss |
|
| esv3573336 | Chr.9:104685959 - 104695251 on Build GRCh38 | Loss |
|
| esv3891705 | Chr.9:104391828 - 106660921 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map