Assay Details
Target Gene Details
Entrez Gene ID: | 54629 |
Gene Name: | family with sequence similarity 63 member B |
Gene Aliases: |
- |
Location: |
Chr.15:58771192-58861900 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM63B | NM_001040450.2 | 1 | 518 | NP_001035540.1 |
| NM_001040453.2 | 1 | 348 | NP_001035543.1 | |
| XM_011521687.1 | 1 | 516 | XP_011519989.1 | |
| XM_011521688.1 | 1 | 516 | XP_011519990.1 | |
| XM_011521689.1 | 1 | 516 | XP_011519991.1 | |
| XM_011521690.1 | 1 | 516 | XP_011519992.1 | |
| XM_017022332.1 | 1 | 516 | XP_016877821.1 | |
| XM_017022333.1 | 1 | 516 | XP_016877822.1 | |
| XM_017022334.1 | 1 | 516 | XP_016877823.1 | |
| XM_017022335.1 | 1 | 516 | XP_016877824.1 | |
| AB075509.1 | 1 | 393 | BAE45756.1 | |
| AK075319.1 | 1 | 396 | BAC11545.1 | |
| AK297291.1 | 1 | 396 | ||
| AL833610.1 | 1 | 519 | ||
| BC140811.1 | 1 | 348 | ||
| BC144616.1 | 1 | 348 | ||
| DA248971.1 | 1 | 516 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34110 | Chr.15:58657792 - 59089435 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map