Assay Details
Target Gene Details
Entrez Gene ID: | 3181 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A2/B1 |
Gene Aliases: |
HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1, HNRPB1, IBMPFD2, RNPA2, SNRPB1 |
Location: |
Chr.7:26189927-26200793 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPA2B1 | NM_002137.3 | NP_002128.1 | ||
| NM_031243.2 | NP_112533.1 | |||
| XM_005249729.1 | XP_005249786.1 | |||
| XM_017012109.1 | XP_016867598.1 | |||
| XM_017012110.1 | XP_016867599.1 | |||
| AK129839.1 | ||||
| AK289429.1 | ||||
| BX537494.1 | 9 | 4066 | ||
| BX538099.1 | ||||
| BX641084.1 | ||||
| M29064.1 | AAA60271.1 | |||
| M29065.1 | AAA36574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891090 | Chr.7:26011300 - 26194682 on Build GRCh38 | Gain |
|
| esv3612615 | Chr.7:25958361 - 26200719 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map