Assay Details
Target Gene Details
Entrez Gene ID: | 8074 |
Gene Name: | fibroblast growth factor 23 |
Gene Aliases: |
ADHR, FGFN, HPDR2, HYPF, PHPTC |
Location: |
Chr.12:4368227-4379728 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF23 | NM_020638.2 | 3 | 2369 | NP_065689.1 |
| AB047858.1 | 3 | 2355 | BAB55889.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss |
|
| nsv557123 | Chr.12:4327466 - 4627403 on Build GRCh38 | Gain |
|
| nsv1040098 | Chr.12:3987637 - 5646161 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map