Assay Details
Target Gene Details
Entrez Gene ID: | 161247 |
Gene Name: | fat storage inducing transmembrane protein 1 |
Gene Aliases: |
FIT1 |
Location: |
Chr.14:24131466-24132849 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FITM1 | NM_203402.2 | 1 | 214 | NP_981947.1 |
| BC139911.1 | 1 | 214 | ||
| BI114004.1 | ||||
| BX347190.2 | 2 | 421 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564057 | Chr.14:24056425 - 24149650 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map