Assay Details
Target Gene Details
Entrez Gene ID: | 347051 |
Gene Name: | solute carrier family 10 member 5 |
Gene Aliases: |
P5 |
Location: |
Chr.8:81693656-81694972 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC10A5 | NM_001010893.2 | 1 | 1017 | NP_001010893.1 |
| AK095808.1 | 1 | 2219 | ||
| AY825924.1 | 1 | 1017 | AAV80707.1 | |
| BC136625.1 | 1 | 1097 | ||
| BC136631.1 | 1 | 1097 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611607 | Chr.8:81405621 - 81768032 on Build GRCh38 | Gain |
|
| nsv831369 | Chr.8:81605833 - 81823586 on Build GRCh38 | Gain+Loss |
|
| nsv831368 | Chr.8:81554926 - 81697395 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map