Assay Details
Target Gene Details
Entrez Gene ID: | 160760 |
Gene Name: | PTC7 protein phosphatase homolog |
Gene Aliases: |
TA-PP2C, TAPP2C |
Location: |
Chr.12:110534432-110583259 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPTC7 | NM_139283.1 | 6 | 1634 | NP_644812.1 |
| AF385435.1 | 6 | 1634 | AAM43836.1 | |
| AK124744.1 | 7 | 1478 | ||
| CR749216.1 | 3 | 756 | CAH18073.1 |
More Information
Set Membership: |
|

Genomic Map