Assay Details
Target Gene Details
Entrez Gene ID: | 51092 |
Gene Name: | SID1 transmembrane family member 2 |
Gene Aliases: |
CGI-40 |
Location: |
Chr.11:117179223-117197445 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIDT2 | NM_001040455.1 | NP_001035545.1 | ||
| AF151799.1 | AAD34036.1 | |||
| AK075137.1 | BAC11427.1 | |||
| AK292501.1 | ||||
| AK292974.1 | ||||
| AK301501.1 | ||||
| AK303023.1 | ||||
| AY358442.1 | AAQ88807.1 | |||
| BC114522.1 | AAI14523.1 | |||
| BC114959.1 | AAI14960.1 | |||
| BX648437.1 | ||||
| KC109742.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2186n54 | Chr.11:117188631 - 117257555 on Build GRCh38 | Loss |
|
| nsv518908 | Chr.11:117180449 - 117195974 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map