Assay Details
Target Gene Details
Entrez Gene ID: | 55034 |
Gene Name: | molybdenum cofactor sulfurase |
Gene Aliases: |
HMCS, MCS, MOS |
Location: |
Chr.18:36187517-36268722 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOCOS | NM_017947.2 | NP_060417.2 | ||
| AK000740.1 | BAA91353.1 | |||
| AK222886.1 | BAD96606.1 | |||
| BC012079.1 | AAH12079.1 | |||
| BQ931455.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv987550 | Chr.18:36187242 - 36197033 on Build GRCh38 | Deletion |
|
| nsv130568 | Chr.18:36193329 - 36201663 on Build GRCh38 | Deletion |
|
| nsv576651 | Chr.18:36017567 - 36959615 on Build GRCh38 | Loss |
|
| nsv522803 | Chr.18:36187786 - 36199742 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map