Assay Details
Target Gene Details
Entrez Gene ID: | 343637 |
Gene Name: | R-spondin 4 |
Gene Aliases: |
C20orf182, CRISTIN4 |
Location: |
Chr.20:958452-1002264 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSPO4 | NM_001029871.3 | 5 | 2307 | NP_001025042.2 |
| NM_001040007.2 | 4 | 2121 | NP_001035096.1 | |
| AK122609.1 | 5 | 2304 | ||
| BQ185331.1 | 1 | 419 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055853 | Chr.20:921359 - 1034331 on Build GRCh38 | Gain |
|
| esv3644952 | Chr.20:386013 - 1119183 on Build GRCh38 | Gain |
|
| nsv1067403 | Chr.20:925142 - 977237 on Build GRCh38 | Gain |
|
| esv2722039 | Chr.20:958895 - 959534 on Build GRCh38 | Deletion |
|
| esv3644966 | Chr.20:887288 - 978010 on Build GRCh38 | Gain |
|
| esv3575346 | Chr.20:884122 - 1168539 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map