Assay Details
Target Gene Details
Entrez Gene ID: | 55584 |
Gene Name: | cholinergic receptor nicotinic alpha 9 subunit |
Gene Aliases: |
HSA243342, NACHRA9 |
Location: |
Chr.4:40335329-40355217 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA9 | NM_017581.3 | 5 | 1628 | NP_060051.2 |
| AJ243342.1 | 5 | 1505 | CAB65091.1 | |
| BC113549.1 | ||||
| BC113575.1 | ||||
| BG218331.1 | 1 | 665 |
More Information
Set Membership: |
|

Genomic Map