Assay Details
Target Gene Details
Entrez Gene ID: | 51053 |
Gene Name: | geminin, DNA replication inhibitor |
Gene Aliases: |
Gem, MGORS6 |
Location: |
Chr.6:24774931-24786099 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMNN | NM_001251989.1 | NP_001238918.1 | ||
| NM_001251990.1 | NP_001238919.1 | |||
| NM_001251991.1 | NP_001238920.1 | |||
| NM_015895.4 | NP_056979.1 | |||
| XM_005249159.1 | XP_005249216.1 | |||
| XM_011514651.2 | XP_011512953.1 | |||
| AF067855.1 | AAC39787.1 | |||
| AK021685.1 | ||||
| AK222992.1 | BAD96712.1 | |||
| AK315743.1 | ||||
| AL558330.3 | ||||
| BC005185.1 | AAH05185.1 | |||
| BC005389.1 | AAH05389.1 | |||
| BG777134.1 | ||||
| BM699599.1 | ||||
| CB131959.1 | ||||
| CB992796.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5227 | Chr.6:24773921 - 24823784 on Build GRCh38 | Deletion |
|
| nsv830609 | Chr.6:24711858 - 24896419 on Build GRCh38 | Gain |
|
| nsv508396 | Chr.6:24779420 - 24846081 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map