Assay Details
Target Gene Details
Entrez Gene ID: | 1592 |
Gene Name: | cytochrome P450 family 26 subfamily A member 1 |
Gene Aliases: |
CP26, CYP26, P450RAI, P450RAI1 |
Location: |
Chr.10:93073475-93077884 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP26A1 | NM_000783.3 | 5 | 970 | NP_000774.2 |
| NM_057157.2 | 5 | 1096 | NP_476498.1 | |
| AF005418.1 | 5 | 963 | AAB88881.1 | |
| AK027560.1 | 5 | 1097 | ||
| AK075374.1 | 5 | 969 |
More Information
Set Membership: |
|

Genomic Map