Assay Details
Target Gene Details
Entrez Gene ID: | 55716 |
Gene Name: | limb development membrane protein 1 like |
Gene Aliases: |
LIMR |
Location: |
Chr.12:49096551-49111535 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
More Information
Set Membership: |
|

Genomic Map