Assay Details
Target Gene Details
Entrez Gene ID: | 25852 |
Gene Name: | armadillo repeat containing 8 |
Gene Aliases: |
GID5, HSPC056, S863-2, VID28 |
Location: |
Chr.3:138187248-138298389 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMC8 | NM_001267041.2 | 1 | 151 | NP_001253970.1 |
| NM_001267042.2 | 1 | 151 | NP_001253971.1 | |
| NM_001282342.1 | 1 | 151 | NP_001269271.1 | |
| NM_014154.4 | 1 | 151 | NP_054873.2 | |
| NM_015396.5 | 1 | 151 | NP_056211.2 | |
| NM_213654.3 | 1 | 151 | NP_998819.1 | |
| XM_006713564.3 | 1 | 146 | XP_006713627.1 | |
| XM_006713565.2 | 1 | 57 | XP_006713628.1 | |
| XM_006713566.3 | 1 | 118 | XP_006713629.1 | |
| XM_006713567.3 | 1 | 146 | XP_006713630.1 | |
| XM_011512632.1 | 1 | 95 | XP_011510934.1 | |
| XM_011512633.1 | 1 | 95 | XP_011510935.1 | |
| XM_017006095.1 | 1 | 73 | XP_016861584.1 | |
| XM_017006096.1 | 1 | 118 | XP_016861585.1 | |
| XM_017006097.1 | 1 | 96 | XP_016861586.1 | |
| XM_017006098.1 | 1 | 118 | XP_016861587.1 | |
| XM_017006099.1 | 1 | 79 | XP_016861588.1 | |
| XM_017006100.1 | 1 | 146 | XP_016861589.1 | |
| XM_017006101.1 | 1 | 73 | XP_016861590.1 | |
| BC032661.1 | AAH32661.1 | |||
| BF966252.1 | 1 | 104 | ||
| DC304503.1 | 1 | 128 | ||
| DC342329.1 | 1 | 116 | ||
| HY004298.1 | 1 | 152 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597911 | Chr.3:138130790 - 138220069 on Build GRCh38 | Gain |
|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
| esv3597913 | Chr.3:138161103 - 138213569 on Build GRCh38 | Gain |
|
| esv2762347 | Chr.3:138132356 - 138191165 on Build GRCh38 | Gain |
|
| esv2759180 | Chr.3:137892947 - 138214993 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map