Assay Details
Target Gene Details
Entrez Gene ID: | 6629 |
Gene Name: | small nuclear ribonucleoprotein polypeptide B2 |
Gene Aliases: |
Msl1, U2B'' |
Location: |
Chr.20:16729964-16741772 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRPB2 | NM_003092.4 | NP_003083.1 | ||
| NM_198220.2 | NP_937863.1 | |||
| AB451234.1 | ||||
| AB451359.1 | ||||
| AK313004.1 | ||||
| BC018022.1 | AAH18022.1 | |||
| BC036737.2 | AAH36737.1 | |||
| DA088017.1 | ||||
| M15841.1 | AAA36796.1 |
More Information
Set Membership: |
|

Genomic Map