Assay Details
Target Gene Details
Entrez Gene ID: | 163933 |
Gene Name: | family with sequence similarity 43 member B |
Gene Aliases: |
- |
Location: |
Chr.1:20552439-20555020 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM43B | NM_207334.2 | 1 | 2049 | NP_997217.1 |
| AK126900.1 | 1 | 2049 | BAC86742.1 | |
| BC142610.1 | 1 | 1692 | ||
| BC142686.1 | 1 | 1940 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map