Assay Details
Target Gene Details
Entrez Gene ID: | 1398 |
Gene Name: | CRK proto-oncogene, adaptor protein |
Gene Aliases: |
CRKII, p38 |
Location: |
Chr.17:1421353-1456267 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRK | NM_005206.4 | 2 | 517 | NP_005197.3 |
| NM_016823.3 | 2 | 517 | NP_058431.2 | |
| AK291060.1 | 2 | 482 | ||
| AU125411.1 | 2 | 517 | ||
| BC001718.1 | 2 | 469 | AAH01718.1 | |
| BC008506.1 | 2 | 500 | AAH08506.1 | |
| BC009837.1 | 2 | 482 | AAH09837.1 | |
| BT007277.1 | 2 | 367 | AAP35941.1 | |
| D10656.1 | 2 | 472 | BAA01505.1 | |
| KU177996.1 | 2 | 367 | ||
| KU177997.1 | 2 | 367 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833334 | Chr.17:1411327 - 1582381 on Build GRCh38 | Loss |
|
| nsv470567 | Chr.17:1391192 - 1463828 on Build GRCh38 | Gain |
|
| nsv1057782 | Chr.17:1351585 - 1512616 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map