Assay Details
Target Gene Details
Entrez Gene ID: | 9050 |
Gene Name: | proline-serine-threonine phosphatase interacting protein 2 |
Gene Aliases: |
MAYP |
Location: |
Chr.18:45983536-46072327 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSTPIP2 | NM_024430.3 | 15 | 1793 | NP_077748.3 |
| XM_011526253.2 | 14 | 1758 | XP_011524555.1 | |
| XM_011526254.1 | 14 | 1670 | XP_011524556.1 | |
| XM_011526256.1 | 14 | 1669 | XP_011524558.1 | |
| AK023100.1 | 11 | 1361 | BAB14404.1 | |
| BC035395.1 | 14 | 1669 | AAH35395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3642419 | Chr.18:45979735 - 45993208 on Build GRCh38 | Loss |
|
| esv3642418 | Chr.18:45974869 - 45987813 on Build GRCh38 | Gain |
|
| nsv2277 | Chr.18:45963749 - 46008613 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map