Assay Details
Target Gene Details
Entrez Gene ID: | 285172 |
Gene Name: | family with sequence similarity 126 member B |
Gene Aliases: |
HYCC2 |
Location: |
Chr.2:200973718-201071671 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM126B | NM_001321618.1 | 11 | 2932 | NP_001308547.1 |
| NM_001321619.1 | 12 | 3143 | NP_001308548.1 | |
| NM_001321621.1 | 11 | 3045 | NP_001308550.1 | |
| NM_001321622.1 | 13 | 3261 | NP_001308551.1 | |
| NM_001321623.1 | 13 | 3198 | NP_001308552.1 | |
| NM_001321624.1 | 12 | 3100 | NP_001308553.1 | |
| NM_001321625.1 | 13 | 3311 | NP_001308554.1 | |
| NM_001321626.1 | 13 | 3331 | NP_001308555.1 | |
| NM_001321627.1 | 14 | 3254 | NP_001308556.1 | |
| NM_001321628.1 | 14 | 3367 | NP_001308557.1 | |
| NM_001321629.1 | 13 | 3086 | NP_001308558.1 | |
| NM_173822.4 | 12 | 3030 | NP_776183.1 | |
| XM_017003879.1 | 14 | 3409 | XP_016859368.1 | |
| XM_017003881.1 | 12 | 3091 | XP_016859370.1 | |
| XM_017003882.1 | 14 | 3310 | XP_016859371.1 | |
| XM_017003883.1 | 13 | 3485 | XP_016859372.1 | |
| AK001843.1 | 1 | 463 | ||
| AK027168.1 | 1 | 905 | ||
| AK097111.1 | 5 | 2273 | ||
| AL832234.1 | 11 | 2889 | ||
| AL832346.1 | 14 | 3249 | ||
| AL833403.1 | 12 | 3014 | ||
| BC039295.1 | 12 | 3028 | AAH39295.1 |
More Information
Set Membership: |
|

Genomic Map