Assay Details
Target Gene Details
Entrez Gene ID: | 1793 |
Gene Name: | dedicator of cytokinesis 1 |
Gene Aliases: |
DOCK180, ced5 |
Location: |
Chr.10:126905409-127452517 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK1 | NM_001290223.1 | NP_001277152.1 | ||
| NM_001380.4 | NP_001371.1 | |||
| XM_011539422.2 | XP_011537724.1 | |||
| XM_011539424.1 | XP_011537726.1 | |||
| XM_017015813.1 | XP_016871302.1 | |||
| XM_017015814.1 | XP_016871303.1 | |||
| XM_017015815.1 | XP_016871304.1 | |||
| XM_017015816.1 | XP_016871305.1 | |||
| BC146857.1 | ||||
| D50857.1 | BAA09454.1 |
Target Gene Details
Entrez Gene ID: | 642938 |
Gene Name: | family with sequence similarity 196 member A |
Gene Aliases: |
C10orf141 |
Location: |
Chr.10:127135426-127196522 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM196A | NM_001039762.2 | 4 | 882 | NP_001034851.1 |
| XM_005252694.3 | 2 | 857 | XP_005252751.1 | |
| XM_017016537.1 | 2 | 4453 | XP_016872026.1 | |
| XM_017016538.1 | 3 | 3869 | XP_016872027.1 | |
| XM_017016539.1 | 2 | 1596 | XP_016872028.1 | |
| XM_017016540.1 | 3 | 1722 | XP_016872029.1 | |
| XM_017016541.1 | 2 | 1158 | XP_016872030.1 | |
| XM_017016542.1 | 3 | 3977 | XP_016872031.1 | |
| XM_017016543.1 | 3 | 682 | XP_016872032.1 | |
| AK127465.1 | 4 | 882 | BAC86991.1 | |
| BC131825.1 | 1 | 367 | ||
| BC137112.1 | 2 | 464 | ||
| BC144478.1 | 2 | 464 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045509 | Chr.10:126162634 - 127193365 on Build GRCh38 | Gain |
|
| esv2759789 | Chr.10:127110261 - 127440758 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map