Assay Details
Target Gene Details
Entrez Gene ID: | 127294 |
Gene Name: | myomesin 3 |
Gene Aliases: |
- |
Location: |
Chr.1:24056041-24112175 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOM3 | NM_152372.3 | 37 | 4281 | NP_689585.3 |
| AF086254.1 | 6 | 371 | ||
| AK093280.1 | 10 | 1167 | BAC04119.1 | |
| AK128233.1 | 29 | 3341 | BAC87343.1 | |
| BC014671.1 | 11 | 801 | AAH14671.1 | |
| BX537911.1 | 36 | 4005 | CAD97895.1 | |
| BX647453.1 | 37 | 4242 | CAH10377.1 | |
| BX647455.1 | 37 | 4281 | CAH10376.1 | |
| BX647644.1 | 37 | 4242 |
More Information
Set Membership: |
|

Genomic Map