Assay Details
Target Gene Details
Entrez Gene ID: | 22909 |
Gene Name: | FANCD2/FANCI-associated nuclease 1 |
Gene Aliases: |
KIAA1018, KMIN, MTMR15, hFAN1 |
Location: |
Chr.15:30903695-30943108 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAN1 | NM_001146094.1 | 2 | 1349 | NP_001139566.1 |
| NM_001146095.1 | 2 | 1246 | NP_001139567.1 | |
| NM_001146096.1 | 2 | 1362 | NP_001139568.1 | |
| NM_014967.4 | 2 | 1349 | NP_055782.3 | |
| XM_005254232.4 | 2 | 1357 | XP_005254289.1 | |
| XM_005254234.4 | 2 | 1403 | XP_005254291.1 | |
| XM_005254235.3 | 2 | 1239 | XP_005254292.1 | |
| XM_011521370.2 | XP_011519672.1 | |||
| XM_011521371.1 | 2 | 1334 | XP_011519673.1 | |
| XM_011521372.2 | 2 | 1353 | XP_011519674.1 | |
| XM_017022012.1 | 2 | 1321 | XP_016877501.1 | |
| XM_017022013.1 | 2 | 1334 | XP_016877502.1 | |
| AB023235.2 | 2 | 1296 | ||
| AK290987.1 | 2 | 1329 | ||
| BC047882.1 | 2 | 1246 | AAH47882.1 | |
| BX647391.1 | 2 | 1343 | ||
| CR936727.1 | 2 | 1329 | ||
| CR936734.1 | 2 | 1319 | ||
| HM560957.1 | 1 | 1058 | ||
| KJ534972.1 | 1 | 1058 | ||
| KJ534973.1 | 1 | 1058 | ||
| KJ535046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832951 | Chr.15:30839036 - 31012266 on Build GRCh38 | Loss |
|
| nsv525795 | Chr.15:30644082 - 31032328 on Build GRCh38 | Gain |
|
| esv3569376 | Chr.15:30781533 - 30987140 on Build GRCh38 | Gain |
|
| esv33507 | Chr.15:30891121 - 31178820 on Build GRCh38 | Loss |
|
| nsv521842 | Chr.15:30848497 - 30980467 on Build GRCh38 | Gain |
|
| nsv1046874 | Chr.15:30715930 - 31002451 on Build GRCh38 | Gain |
|
| esv3636036 | Chr.15:30818302 - 31038636 on Build GRCh38 | Gain |
|
| esv3892649 | Chr.15:30905361 - 30918448 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map