Assay Details
Target Gene Details
Entrez Gene ID: | 83854 |
Gene Name: | angiopoietin like 6 |
Gene Aliases: |
AGF, ARP5 |
Location: |
Chr.19:10092337-10106194 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPTL6 | NM_001321411.1 | NP_001308340.1 | ||
| NM_031917.2 | NP_114123.2 | |||
| XM_005260091.3 | XP_005260148.1 | |||
| XM_011528347.2 | XP_011526649.1 | |||
| XM_011528348.2 | XP_011526650.1 | |||
| XM_011528349.2 | XP_011526651.1 | |||
| XM_011528350.2 | XP_011526652.1 | |||
| XM_017027347.1 | XP_016882836.1 | |||
| AB054064.1 | BAB91248.1 | |||
| AF230330.1 | AAK06404.1 | |||
| AY358276.1 | AAQ88643.1 | |||
| BC142632.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2414 | Chr.19:10063769 - 10097402 on Build GRCh38 | Insertion |
|
| nsv458355 | Chr.19:10061840 - 10102478 on Build GRCh38 | Loss |
|
| nsv833743 | Chr.19:10077649 - 10251913 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs147658769] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map