Assay Details
Target Gene Details
Entrez Gene ID: | 23596 |
Gene Name: | opsin 3 |
Gene Aliases: |
ECPN, PPP1R116 |
Location: |
Chr.1:241593150-241640399 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPN3 | NM_014322.2 | 3 | 1007 | NP_055137.2 |
| AF140242.1 | 3 | 917 | AAD32671.1 | |
| AF303588.1 | 3 | 964 | AAK37447.1 | |
| AK310307.1 | 3 | 490 | ||
| BC036773.1 | 3 | 959 | AAH36773.1 | |
| BC062622.1 | 3 | 788 | AAH62622.1 | |
| BC073872.1 | 2 | 612 | AAH73872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549460 | Chr.1:241331278 - 241669985 on Build GRCh38 | Gain |
|
| nsv1002214 | Chr.1:241295639 - 241936488 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map