Assay Details
Target Gene Details
Entrez Gene ID: | 26470 |
Gene Name: | seizure related 6 homolog like 2 |
Gene Aliases: |
BSRPA, PSK-1 |
Location: |
Chr.16:29871159-29899573 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEZ6L2 | NM_001114099.2 | 6 | 1018 | NP_001107571.1 |
| NM_001114100.2 | 4 | 886 | NP_001107572.1 | |
| NM_001243332.1 | 6 | 1228 | NP_001230261.1 | |
| NM_001243333.1 | 5 | 1096 | NP_001230262.1 | |
| NM_012410.3 | 6 | 1018 | NP_036542.1 | |
| NM_201575.3 | 6 | 1228 | NP_963869.2 | |
| XM_005255252.2 | 6 | 1228 | XP_005255309.1 | |
| XM_017023135.1 | 4 | 985 | XP_016878624.1 | |
| AB105376.1 | 6 | 857 | ||
| AJ245820.1 | 6 | 857 | CAB57950.1 | |
| AJ245821.1 | 6 | 857 | CAB57951.1 | |
| AJ245822.1 | 6 | 857 | CAB57952.1 | |
| AK027407.1 | 4 | 498 | ||
| AK027676.1 | 3 | 468 | ||
| AK074998.1 | 2 | 186 | BAC11345.1 | |
| AK293675.1 | 5 | 941 | ||
| AK299103.1 | 6 | 1225 | ||
| AY358404.1 | 6 | 1217 | AAQ88770.1 | |
| BC000567.2 | 4 | 686 | AAH00567.1 | |
| EL733582.1 | ||||
| KJ534938.1 | 6 | 983 | ||
| KJ534939.1 | 4 | 647 | ||
| KJ534940.1 | 6 | 983 | ||
| KJ534941.1 | 4 | 641 | ||
| KJ534988.1 | 6 | 989 | ||
| KJ534989.1 | ||||
| KJ535068.1 | 6 | 894 | ||
| KJ535092.1 | 6 | 982 |

Genomic Map