Assay Details
Target Gene Details
Entrez Gene ID: | 2526 |
Gene Name: | fucosyltransferase 4 |
Gene Aliases: |
CD15, ELFT, FCT3A, FUC-TIV, FUTIV, LeX, SSEA-1 |
Location: |
Chr.11:94543851-94549898 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FUT4 | NM_002033.3 | 1 | 1805 | NP_002024.1 |
| BC136373.1 | 1 | 1685 | ||
| BC136374.1 | 1 | 1685 | ||
| M58596.1 | 1 | 1204 | AAA63172.1 | |
| M58597.1 | 1 | 1695 | AAA63173.1 |
Target Gene Details
Entrez Gene ID: | 105369438 |
Gene Name: | uncharacterized LOC105369438 |
Gene Aliases: |
- |
Location: |
Chr.11:94545332-94740355 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105369438 | NR_135093.1 | 5 | 715 | |
| BI522712.1 | 5 | 628 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580028 | Chr.11:94534020 - 94546921 on Build GRCh38 | Gain |
|
| nsv951059 | Chr.11:94545635 - 94552434 on Build GRCh38 | Duplication |
|
| esv2671948 | Chr.11:94349534 - 94645424 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map