Assay Details
Target Gene Details
Entrez Gene ID: | 163351 |
Gene Name: | guanylate binding protein family member 6 |
Gene Aliases: |
- |
Location: |
Chr.1:89363877-89388160 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP6 | NM_001320257.1 | 8 | 1461 | NP_001307186.1 |
| NM_198460.2 | 10 | 1802 | NP_940862.2 | |
| XM_011540835.2 | 10 | 1631 | XP_011539137.1 | |
| AI199897.1 | ||||
| AK131329.1 | 5 | 684 | BAD18489.1 | |
| AK131356.1 | 10 | 1786 | BAD18509.1 | |
| AK290303.1 | 10 | 1622 | ||
| AK299408.1 | 9 | 1437 | ||
| AK299466.1 | 8 | 1278 | ||
| AK303987.1 | 10 | 1546 | ||
| AK315834.1 | 10 | 1620 | ||
| BC131713.1 | 10 | 1580 | ||
| BX537949.1 | 10 | 1621 | CAD97917.1 | |
| BX647907.1 | 10 | 1619 |
More Information
Set Membership: |
|

Genomic Map