Assay Details
Target Gene Details
Entrez Gene ID: | 3141 |
Gene Name: | holocarboxylase synthetase |
Gene Aliases: |
HCS |
Location: |
Chr.21:36750888-36990254 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HLCS | NM_000411.6 | 12 | 2903 | NP_000402.3 |
| NM_001242784.1 | 12 | 3837 | NP_001229713.1 | |
| NM_001242785.1 | 12 | 2996 | NP_001229714.1 | |
| XM_005260953.3 | 11 | 2865 | XP_005261010.1 | |
| XM_005260955.3 | 11 | 2736 | XP_005261012.1 | |
| XM_005260956.3 | 11 | 2912 | XP_005261013.1 | |
| XM_006723994.2 | 11 | 2850 | XP_006724057.1 | |
| XM_006723995.1 | 12 | 3028 | XP_006724058.1 | |
| XM_011529538.1 | 12 | 2883 | XP_011527840.1 | |
| XM_011529539.2 | 12 | 2888 | XP_011527841.1 | |
| XM_011529541.2 | 11 | 2739 | XP_011527843.1 | |
| XM_017028330.1 | 11 | 2686 | XP_016883819.1 | |
| D23672.2 | 12 | 2894 | BAA04902.1 | |
| D87328.1 | 12 | 3654 | BAA13332.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73398122] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map