Assay Details
Target Gene Details
Entrez Gene ID: | 7415 |
Gene Name: | valosin containing protein |
Gene Aliases: |
ALS14, CMT2Y, HEL-220, HEL-S-70, IBMPFD, IBMPFD1, TERA, p97 |
Location: |
Chr.9:35056068-35072742 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VCP | NM_007126.3 | NP_009057.1 | ||
| AF100752.1 | AAD43016.1 | |||
| AK307735.1 | ||||
| AK312310.1 | ||||
| AL137377.1 | CAB70717.1 | |||
| BC007562.1 | AAH07562.2 | |||
| BC110913.1 | AAI10914.1 | |||
| BC121794.1 | ||||
| BC122550.1 | ||||
| FJ224344.1 | ||||
| FJ224352.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620367 | Chr.9:35058196 - 35071684 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map