Assay Details
Target Gene Details
Entrez Gene ID: | 7134 |
Gene Name: | troponin C1, slow skeletal and cardiac type |
Gene Aliases: |
CMD1Z, CMH13, TN-C, TNC, TNNC |
Location: |
Chr.3:52451091-52454041 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNNC1 | NM_003280.2 | NP_003271.1 | ||
| AF020769.1 | AAB91994.1 | |||
| AK313743.1 | ||||
| BC030244.1 | AAH30244.1 | |||
| CR541865.1 | CAG46663.1 | |||
| CR541885.1 | CAG46683.1 | |||
| X07897.1 | CAA30736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834697 | Chr.3:52366562 - 52545401 on Build GRCh38 | Loss |
|
| nsv954860 | Chr.3:52439285 - 52456384 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map