Assay Details
Target Gene Details
Entrez Gene ID: | 10326 |
Gene Name: | signal regulatory protein beta 1 |
Gene Aliases: |
CD172b, SIRP-BETA-1 |
Location: |
Chr.20:1564193-1620457 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRPB1 | XM_005260641.3 | 6 | 1732 | XP_005260698.1 |
| XM_005260643.3 | 5 | 1399 | XP_005260700.1 | |
| XM_005260644.3 | 4 | 1082 | XP_005260701.1 | |
| XM_017027577.1 | 6 | 2195 | XP_016883066.1 | |
| XM_017027581.1 | 7 | 2040 | XP_016883070.1 | |
| XM_017027582.1 | 7 | 2080 | XP_016883071.1 | |
| XM_017027585.1 | 5 | 1832 | XP_016883074.1 | |
| BC025286.1 | 4 | 1072 | AAH25286.1 | |
| BC075835.1 | 4 | 1072 | AAH75835.1 | |
| BC131747.1 | 4 | 1056 | ||
| BP268958.1 | ||||
| Y10376.1 | 6 | 1711 | CAA71404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2424715 | Chr.20:1564995 - 1565893 on Build GRCh38 | Insertion |
|
| nsv9775 | Chr.20:1554339 - 1583014 on Build GRCh38 | Loss |
|
| nsv522008 | Chr.20:1359746 - 1657932 on Build GRCh38 | Gain |
|
| nsv1160672 | Chr.20:1564764 - 1580934 on Build GRCh38 | Deletion |
|
| nsv510779 | Chr.20:1528684 - 1633316 on Build GRCh38 | Deletion |
|
| esv2758777 | Chr.20:1496135 - 1698610 on Build GRCh38 | Gain+Loss |
|
| nsv585206 | Chr.20:1405829 - 1733716 on Build GRCh38 | Gain |
|
| esv33848 | Chr.20:1565055 - 1570417 on Build GRCh38 | Loss |
|
| nsv833894 | Chr.20:1538787 - 1772720 on Build GRCh38 | Gain |
|
| esv3644980 | Chr.20:1509522 - 1569123 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map