Assay Details
Target Gene Details
Entrez Gene ID: | 23582 |
Gene Name: | cyclin D1 binding protein 1 |
Gene Aliases: |
DIP1, GCIP, HHM |
Location: |
Chr.15:43185118-43197177 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNDBP1 | NM_012142.4 | 8 | 1008 | NP_036274.3 |
| NR_027513.2 | 6 | 831 | ||
| NR_027514.2 | 7 | 911 | ||
| NR_045998.1 | 7 | 928 | ||
| NR_045999.1 | 5 | 771 | ||
| XM_006720448.1 | 6 | 930 | XP_006720511.1 | |
| AF082569.1 | 8 | 867 | AAD11777.1 | |
| AF087852.1 | 8 | 853 | AAP97163.1 | |
| AF113535.1 | 6 | 817 | AAF14872.1 | |
| AF132034.1 | 8 | 823 | AAF77613.1 | |
| AF246144.1 | 8 | 825 | AAF67182.1 | |
| AK075146.1 | 8 | 837 | BAC11433.1 | |
| AK075296.1 | 6 | 822 | BAC11530.1 | |
| AK124193.1 | 3 | 1412 | ||
| AK128849.1 | 4 | 2052 | BAC87645.1 | |
| AK225143.1 | 8 | 823 | ||
| AK225645.1 | 8 | 837 | ||
| AK290665.1 | 8 | 837 | ||
| AK290707.1 | 7 | 790 | ||
| AK295017.1 | 7 | 740 | ||
| AL552334.3 | 8 | 947 | ||
| BC009689.1 | 8 | 851 | AAH09689.1 | |
| BQ439439.1 | 5 | 585 | ||
| CR450331.1 | 8 | 777 | CAG29327.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569241 | Chr.15:42908911 - 43230724 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map