Assay Details
Target Gene Details
Entrez Gene ID: | 5900 |
Gene Name: | ral guanine nucleotide dissociation stimulator |
Gene Aliases: |
RGDS, RGF, RalGEF |
Location: |
Chr.9:133097720-133149220 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RALGDS | NM_001042368.2 | 18 | 3276 | NP_001035827.1 |
| NM_001271774.1 | 18 | 3188 | NP_001258703.1 | |
| NM_001271775.1 | 18 | 3281 | NP_001258704.1 | |
| NM_001271776.1 | 18 | 3261 | NP_001258705.1 | |
| NM_006266.3 | 18 | 3297 | NP_006257.1 | |
| AB037729.1 | 11 | 5406 | BAA92546.1 | |
| AF027169.1 | 18 | 3192 | AAQ13414.1 | |
| AK000242.1 | 13 | 2254 | ||
| AK024575.1 | 10 | 1851 | ||
| AK056462.1 | 17 | 2974 | ||
| AK074114.1 | 18 | 4193 | BAB84940.1 | |
| AK090450.1 | 12 | 5265 | BAC03431.1 | |
| AK127524.1 | 18 | 3281 | BAC87018.1 | |
| BC021581.1 | 6 | 1373 | AAH21581.1 | |
| BC033198.1 | 10 | 1693 | AAH33198.1 | |
| BC059362.1 | 18 | 3251 | AAH59362.1 | |
| BM805052.1 | 3 | 862 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| dgv12929n54 | Chr.9:133090975 - 133155959 on Build GRCh38 | Loss |
|
| nsv951797 | Chr.9:133097514 - 133173313 on Build GRCh38 | Deletion |
|
| nsv831744 | Chr.9:132980050 - 133174043 on Build GRCh38 | Gain+Loss |
|
| dgv12928n54 | Chr.9:133078923 - 133167883 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map