Assay Details
Target Gene Details
Entrez Gene ID: | 4938 |
Gene Name: | 2'-5'-oligoadenylate synthetase 1 |
Gene Aliases: |
E18/E16, IFI-4, OIAS, OIASI |
Location: |
Chr.12:112906777-112932190 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OAS1 | NM_001032409.2 | NP_001027581.1 | ||
| NM_001320151.1 | NP_001307080.1 | |||
| NM_016816.3 | 6 | 1400 | NP_058132.2 | |
| XM_006719434.1 | 6 | 2019 | XP_006719497.1 | |
| XM_011538413.1 | XP_011536715.1 | |||
| XM_017019361.1 | 6 | 1203 | XP_016874850.1 | |
| XM_017019362.1 | 6 | 1052 | XP_016874851.1 | |
| AA085948.1 | ||||
| AJ629455.1 | CAF33358.1 | |||
| AK225384.1 | 6 | 1200 | ||
| AY730627.1 | 6 | 1145 | AAW63049.1 | |
| AY730628.1 | AAW63050.1 | |||
| BF055326.1 | 1 | 431 | ||
| DQ823040.1 | ||||
| DQ914956.1 | ||||
| X02661.1 | 3 | 447 | CAA26497.1 | |
| X02875.1 | 3 | 270 | CAA26634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832515 | Chr.12:112778024 - 112926931 on Build GRCh38 | Gain |
|
| nsv1047747 | Chr.12:112917435 - 113003116 on Build GRCh38 | Gain |
|
| nsv1042636 | Chr.12:112627854 - 113014974 on Build GRCh38 | Gain |
|
| nsv832516 | Chr.12:112883300 - 113040797 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map