Assay Details
Target Gene Details
Entrez Gene ID: | 23386 |
Gene Name: | NudC domain containing 3 |
Gene Aliases: |
NudCL |
Location: |
Chr.7:44382366-44490875 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDCD3 | NM_015332.3 | 6 | 1514 | NP_056147.2 |
| XM_011515247.2 | XP_011513549.1 | |||
| XM_017011909.1 | 7 | 1540 | XP_016867398.1 | |
| XM_017011910.1 | 7 | 1398 | XP_016867399.1 | |
| AB028991.1 | 6 | 1308 | BAA83020.1 | |
| AK024226.1 | 5 | 1022 | BAB14855.1 | |
| AK026126.1 | 1 | 309 | ||
| AK124729.1 | 1 | 1417 | ||
| AK307613.1 | 4 | 821 | ||
| BC003691.2 | 6 | 1375 | AAH03691.1 | |
| BC011673.2 | 5 | 1134 | ||
| BC035014.2 | 6 | 1412 | AAH35014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526063 | Chr.7:44338308 - 44409902 on Build GRCh38 | Loss |
|
| nsv830984 | Chr.7:44292498 - 44482587 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map