Assay Details
Target Gene Details
Entrez Gene ID: | 10994 |
Gene Name: | ilvB acetolactate synthase like |
Gene Aliases: |
209L8, AHAS, HACL1L, ILV2H |
Location: |
Chr.19:15114974-15125799 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ILVBL | NM_006844.4 | NP_006835.2 | ||
| XM_005259717.4 | XP_005259774.1 | |||
| XM_011527651.2 | XP_011525953.1 | |||
| AB209065.1 | 7 | 2986 | BAD92302.1 | |
| BC000109.1 | AAH00109.1 | |||
| BC011722.2 | AAH11722.1 | |||
| BC011761.2 | AAH11761.1 | |||
| U61263.1 | AAC50934.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9668 | Chr.19:15101800 - 15131511 on Build GRCh38 | Gain |
|
| esv34154 | Chr.19:15101623 - 15284812 on Build GRCh38 | Loss |
|
| esv2758746 | Chr.19:14986138 - 15148368 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map