Assay Details
Target Gene Details
Entrez Gene ID: | 200844 |
Gene Name: | chromosome 3 open reading frame 67 |
Gene Aliases: |
- |
Location: |
Chr.3:58716415-59050078 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
Target Gene Details
Entrez Gene ID: | 101929238 |
Gene Name: | C3orf67 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:58824471-59019093 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf67-AS1 | NR_110820.1 | |||
| AK090895.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4749n100 | Chr.3:58504269 - 60219035 on Build GRCh38 | Gain |
|
| esv3596258 | Chr.3:58813251 - 58852994 on Build GRCh38 | Loss |
|
| nsv1002940 | Chr.3:58775707 - 58858302 on Build GRCh38 | Gain |
|
| esv2763270 | Chr.3:58824677 - 58841613 on Build GRCh38 | Loss |
|
| nsv511230 | Chr.3:58814270 - 58831876 on Build GRCh38 | Loss |
|
| esv3893702 | Chr.3:58557785 - 58858605 on Build GRCh38 | Loss |
|
| nsv834704 | Chr.3:58701751 - 58851925 on Build GRCh38 | Loss |
|

Genomic Map