Assay Details
Target Gene Details
Entrez Gene ID: | 59341 |
Gene Name: | transient receptor potential cation channel subfamily V member 4 |
Gene Aliases: |
BCYM3, CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VRL2, VROAC |
Location: |
Chr.12:109783087-109833407 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPV4 | NM_001177428.1 | 5 | 1135 | NP_001170899.1 |
| NM_001177431.1 | 7 | 1143 | NP_001170902.1 | |
| NM_001177433.1 | NP_001170904.1 | |||
| NM_021625.4 | 7 | 1334 | NP_067638.3 | |
| NM_147204.2 | NP_671737.1 | |||
| XM_005253918.1 | 7 | 1694 | XP_005253975.1 | |
| XM_011538630.2 | 7 | 2437 | XP_011536932.2 | |
| XM_011538631.2 | 6 | 2297 | XP_011536933.2 | |
| XM_011538632.2 | XP_011536934.2 | |||
| XM_011538633.2 | XP_011536935.2 | |||
| XM_011538634.2 | 7 | 2439 | XP_011536936.2 | |
| XM_011538635.2 | 7 | 2441 | XP_011536937.1 | |
| XM_011538636.2 | 7 | 2442 | XP_011536938.1 | |
| XM_017019774.1 | 7 | 2067 | XP_016875263.1 | |
| AB032427.1 | 6 | 1340 | BAB69040.1 | |
| AB073669.1 | BAC06573.1 | |||
| AB100308.1 | 6 | 1245 | BAC55864.1 | |
| AF258465.1 | 6 | 1245 | AAG16127.1 | |
| AF263523.1 | 6 | 1245 | AAG28029.1 | |
| AF279673.1 | 6 | 1041 | AAK69487.1 | |
| AJ296305.1 | 7 | 1334 | CAC82937.1 | |
| AL834192.1 | 7 | 1307 | ||
| BC117426.1 | 7 | 1328 | ||
| BC143315.1 | 7 | 1328 | ||
| DQ059644.1 | 5 | 1104 | AAZ04918.1 | |
| DQ059645.1 | 7 | 1143 | AAZ04919.1 | |
| DQ059646.1 | AAZ04920.1 |
More Information
Set Membership: |
|

Genomic Map