Assay Details
Target Gene Details
Entrez Gene ID: | 80778 |
Gene Name: | zinc finger protein 34 |
Gene Aliases: |
KOX32 |
Location: |
Chr.8:144772224-144787371 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF34 | NM_001286769.1 | 6 | 498 | NP_001273698.1 |
| NM_001286770.1 | 5 | 374 | NP_001273699.1 | |
| NM_030580.4 | 6 | 494 | NP_085057.3 | |
| XM_011517314.2 | 6 | 1821 | XP_011515616.1 | |
| XM_011517315.2 | 6 | 738 | XP_011515617.1 | |
| XM_011517316.2 | 7 | 543 | XP_011515618.1 | |
| XM_011517318.2 | 6 | 1821 | XP_011515620.1 | |
| XM_011517320.2 | 5 | 404 | XP_011515622.1 | |
| XM_017013872.1 | 7 | 544 | XP_016869361.1 | |
| XM_017013873.1 | 6 | 484 | XP_016869362.1 | |
| XM_017013874.1 | 6 | 518 | XP_016869363.1 | |
| XM_017013875.1 | 7 | 684 | XP_016869364.1 | |
| XM_017013876.1 | 5 | 358 | XP_016869365.1 | |
| AK096508.1 | 5 | 345 | ||
| AL833814.1 | 6 | 478 | CAD38677.1 | |
| BC004480.2 | 6 | 453 | AAH04480.1 | |
| BC028136.1 | 3 | 596 | AAH28136.1 | |
| BP199961.1 | 5 | 374 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522459 | Chr.8:144519234 - 144845293 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map