Assay Details
Target Gene Details
Entrez Gene ID: | 151636 |
Gene Name: | deltex E3 ubiquitin ligase 3L |
Gene Aliases: |
BBAP |
Location: |
Chr.3:122564338-122575203 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTX3L | NM_138287.3 | 3 | 1411 | NP_612144.1 |
| AF484416.1 | 3 | 1322 | AAL90859.1 | |
| AK125086.1 | 3 | 1411 | ||
| AL833598.1 | 3 | 1344 | ||
| AY225123.1 | 3 | 1456 | AAP57517.1 | |
| AY780792.1 | AAV98362.1 | |||
| BC042191.1 | 3 | 1330 | AAH42191.1 | |
| BC060509.1 | 3 | 1353 | AAH60509.1 | |
| BX648267.1 | 1 | 835 | ||
| BX648645.1 | 3 | 1164 |
More Information
Additional Information:
For this assay, SNP(s) [rs77786095] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map