Assay Details
Target Gene Details
Entrez Gene ID: | 64221 |
Gene Name: | roundabout guidance receptor 3 |
Gene Aliases: |
HGPPS, HGPS, RBIG1, RIG1 |
Location: |
Chr.11:124865409-124881474 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ROBO3 | NM_022370.3 | 2 | 588 | NP_071765.2 |
| XM_017018122.1 | 1 | 353 | XP_016873611.1 | |
| AK291045.1 | 2 | 522 | ||
| AY509035.1 | 2 | 611 | AAS91662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv531 | Chr.11:124866914 - 124904425 on Build GRCh38 | Insertion |
|
| nsv1054824 | Chr.11:124861873 - 124901623 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map