Assay Details
Target Gene Details
Entrez Gene ID: | 4998 |
Gene Name: | origin recognition complex subunit 1 |
Gene Aliases: |
HSORC1, ORC1L, PARC1 |
Location: |
Chr.1:52371408-52404471 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORC1 | NM_001190818.1 | 5 | 897 | NP_001177747.1 |
| NM_001190819.1 | 5 | 900 | NP_001177748.1 | |
| NM_004153.3 | 5 | 900 | NP_004144.2 | |
| XM_011541527.2 | 5 | 792 | XP_011539829.1 | |
| XM_017001388.1 | 5 | 900 | XP_016856877.1 | |
| XM_017001389.1 | 5 | 875 | XP_016856878.1 | |
| AK291909.1 | 5 | 875 | ||
| AK303420.1 | 5 | 841 | ||
| BC011539.1 | 5 | 813 | AAH11539.1 | |
| U40152.1 | 5 | 888 | AAC50325.1 | |
| U43416.1 | 5 | 880 | AAA86260.1 |
More Information
Set Membership: |
|

Genomic Map