Assay Details
Target Gene Details
Entrez Gene ID: | 404266 |
Gene Name: | HOXB cluster antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.17:48590420-48606414 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXB-AS3 | NR_033201.2 | |||
| NR_033202.2 | ||||
| NR_033203.1 | ||||
| NR_033204.2 | ||||
| NR_033205.1 | ||||
| NR_110329.1 | ||||
| NR_110330.1 | ||||
| NR_110331.1 | ||||
| AA299267.1 | ||||
| BF431268.1 | ||||
| BG203124.1 | ||||
| BG203609.1 | ||||
| BG217847.1 | ||||
| CB046509.1 | ||||
| CF127739.1 | ||||
| DA741273.1 | ||||
| W39335.1 |
Target Gene Details
Entrez Gene ID: | 3216 |
Gene Name: | homeobox B6 |
Gene Aliases: |
HOX2, HOX2B, HU-2, Hox-2.2 |
Location: |
Chr.17:48595736-48604992 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXB6 | NM_018952.4 | 3 | 485 | NP_061825.2 |
| XM_005257283.3 | 2 | 727 | XP_005257340.1 | |
| XM_005257284.3 | 2 | 783 | XP_005257341.1 | |
| XM_011524727.2 | 3 | 981 | XP_011523029.1 | |
| AK292200.1 | 2 | 783 | ||
| AW370639.1 | ||||
| BC014651.1 | 3 | 485 | AAH14651.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833471 | Chr.17:48540425 - 48705983 on Build GRCh38 | Gain |
|
| dgv5676n54 | Chr.17:48539657 - 48627174 on Build GRCh38 | Loss |
|
| nsv833469 | Chr.17:48414487 - 48632044 on Build GRCh38 | Loss |
|
| esv2758694 | Chr.17:48525311 - 48759475 on Build GRCh38 | Gain |
|
| dgv369n27 | Chr.17:48543040 - 48621348 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map