Assay Details
Target Gene Details
Entrez Gene ID: | 54704 |
Gene Name: | pyruvate dehyrogenase phosphatase catalytic subunit 1 |
Gene Aliases: |
PDH, PDP, PDPC, PPM2A, PPM2C |
Location: |
Chr.8:93916855-93926068 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDP1 | NM_001161779.1 | 3 | 3482 | NP_001155251.1 |
| NM_001161780.1 | 3 | 3356 | NP_001155252.1 | |
| NM_001161781.1 | 2 | 3343 | NP_001155253.1 | |
| NM_018444.3 | 2 | 3400 | NP_060914.2 | |
| XM_011517135.2 | 3 | 3253 | XP_011515437.1 | |
| XM_011517136.1 | 3 | 3360 | XP_011515438.1 | |
| XM_017013587.1 | 2 | 3415 | XP_016869076.1 | |
| XM_017013588.1 | 3 | 3323 | XP_016869077.1 | |
| AK022057.1 | 1 | 1987 | ||
| AK126862.1 | 1 | 3777 | ||
| BQ422289.1 | 1 | 1008 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1022473 | Chr.8:91854892 - 94250166 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map