Assay Details
Target Gene Details
Entrez Gene ID: | 7088 |
Gene Name: | transducin like enhancer of split 1 |
Gene Aliases: |
ESG, ESG1, GRG1 |
Location: |
Chr.9:81583683-81689535 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv821665 | Chr.9:80752718 - 82292739 on Build GRCh38 | Loss |
|
| nsv831638 | Chr.9:81538144 - 81707769 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map