Assay Details
Target Gene Details
Entrez Gene ID: | 1947 |
Gene Name: | ephrin B1 |
Gene Aliases: |
CFND, CFNS, EFB1, EFL3, EPLG2, Elk-L, LERK2 |
Location: |
Chr.X:68828997-68842164 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFNB1 | NM_004429.4 | 5 | 1567 | NP_004420.1 |
| BC016649.1 | 5 | 1567 | AAH16649.1 | |
| BC052979.1 | 5 | 1586 | AAH52979.1 | |
| L37361.1 | 5 | 1313 | AAA52369.1 | |
| U09303.1 | 5 | 1488 | AAB41127.1 | |
| U09304.1 | 5 | 1094 | AAA53093.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516469 | Chr.X:68640253 - 68949573 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map